Skip to Content
Merck
All Photos(1)

Key Documents

M0944

Sigma-Aldrich

Anti-Msx1 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-HOX7, Anti-msh homeo box homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€506.00

€506.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€506.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

€506.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 30.9 kDa

species reactivity

human, rodent

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSX1(4487)
mouse ... Msx1(17701)
rat ... Msx1(81710)

Immunogen

synthetic peptide corresponding to human Msx1 (amino acids 42-52), conjugated to BSA.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Target description

Msx1 encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minglian Zhao et al.
Molecular and cellular biology, 33(16), 3099-3112 (2013-06-12)
Improving the knowledge of disease-causing genes is a unique challenge in human health. Although it is known that genes causing similar diseases tend to lie close to one another in a network of protein-protein or functional interactions, the identification of
Marcos Simões-Costa et al.
Developmental cell, 34(5), 544-554 (2015-08-11)
Neural crest cells are induced at the neural plate border by the combined action of transcription factors and signaling molecules. Here, we show that Axud1, a downstream effector of Wnt signaling, represents a critical missing link that integrates signaling and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service