Skip to Content
Merck
All Photos(1)

Key Documents

L8288

Sigma-Aldrich

Lappaconitine hydrobromide

≥96%, solid

Synonym(s):

Allapinine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H44N2O8 · HBr
Molecular Weight:
665.61
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥96%

form

solid

storage temp.

2-8°C

SMILES string

Br.CCN1C[C@@]2(CC[C@H](OC)[C@@]34[C@@H]2C[C@@H]([C@@H]13)[C@@]5(O)C[C@H](OC)[C@H]6C[C@@H]4[C@]5(O)[C@H]6OC)OC(=O)c7ccccc7NC(C)=O

InChI

1S/C32H44N2O8.BrH/c1-6-34-16-29(42-28(36)18-9-7-8-10-21(18)33-17(2)35)12-11-25(40-4)31-23(29)14-20(26(31)34)30(37)15-22(39-3)19-13-24(31)32(30,38)27(19)41-5;/h7-10,19-20,22-27,37-38H,6,11-16H2,1-5H3,(H,33,35);1H/t19-,20+,22+,23-,24+,25+,26?,27+,29-,30+,31+,32+;/m1./s1

InChI key

CFFYROOPXPKMEQ-OPLXFBIMSA-N

Biochem/physiol Actions

Selective blocker of the TTX-sensitive Na+ channels, without influence on the activation threshold of Na+ channels.

Warning

Protect from light.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service