Key Documents
L6539
Sigma-Aldrich
Laser Tough-Spots®
diam. 1/2 in., yellow
Synonym(s):
solvent resistant labels, temperature resistant labels
Select a Size
1 PKG
€75.20
€75.20
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
€75.20
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
€75.20
Please contact Customer Service for Availability
material
polyolefin
yellow
packaging
pack of 3840 ea (192 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-2500
diam.
1/2 in.
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)