Skip to Content
Merck
All Photos(1)

Key Documents

L5263

Sigma-Aldrich

Lyticase from Arthrobacter luteus

partially purified powder, ≥1,500 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204

form

partially purified powder

specific activity

≥1,500 units/mg protein

composition

Protein, ≥10% biuret

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lyticase hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan.

Unit Definition

One unit will produce a ΔA800 of 0.001 per min at pH 7.5 at 25 °C, using a suspension of yeast as substrate in a 3 mL reaction mixture.

Physical form

Partially purified powder containing ammonium sulfate and stabilizer

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Physical mapping by pulsed-field gel electrophoresis.
J Maule
Methods in molecular biology (Clifton, N.J.), 68, 93-121 (1997-01-01)
Construction of chromosome-specific libraries of yeast artificial chromosome recombinants from somatic hybrid cell lines.
B Arveiler
Methods in molecular biology (Clifton, N.J.), 29, 379-402 (1994-01-01)
B N Doebbeling et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 16(3), 377-383 (1993-03-01)
Candida species are important nosocomial pathogens, particularly in immunocompromised and critically ill patients. A variety of methods have been used to differentiate strains, but an optimal system has not been established. We compared methods for typing a panel of nine
Sanford J Silverman et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(15), 6946-6951 (2010-03-26)
Oscillations in patterns of expression of a large fraction of yeast genes are associated with the "metabolic cycle," usually seen only in prestarved, continuous cultures of yeast. We used FISH of mRNA in individual cells to test the hypothesis that
Markus M M Bisschops et al.
Biochimica et biophysica acta, 1864(1), 231-242 (2016-11-08)
Non-dividing Saccharomyces cerevisiae cultures are highly relevant for fundamental and applied studies. However, cultivation conditions in which non-dividing cells retain substantial metabolic activity are lacking. Unlike stationary-phase (SP) batch cultures, the current experimental paradigm for non-dividing yeast cultures, cultivation under

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service