Skip to Content
Merck
All Photos(3)

Key Documents

L5014

Sigma-Aldrich

Lipopolysaccharides (rough strains) from Escherichia coli J5 (Rc mutant)

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€258.00
10 MG
€447.00

€258.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€258.00
10 MG
€447.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

€258.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Escherichia coli (J5 Rc mutant)

Quality Level

form

lyophilized powder

impurities

<3% Protein (Lowry)

color

white to faint beige

solubility

water: 0.90-1.10 mg/mL, hazy to turbid, colorless to faintly yellow

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

Prepared by phenol-chloroform-petroleum ether extraction.
The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyu Zhang et al.
Oncology letters, 12(2), 1034-1040 (2016-07-23)
Hypoxia-inducible factor-1α (HIF-1α) and toll-like receptor 4 (TLR4) are involved in numerous mechanisms of cancer biology, including cell proliferation and survival; however the interaction of the two factors under hypoxic conditions remains unclear. The present study investigated the in vitro
M Nashleanas et al.
Infection and immunity, 68(3), 1428-1434 (2000-02-26)
The ability to activate macrophages in vitro for nitric oxide production and killing of Leishmania major parasites is dependent on tumor necrosis factor, although L. major-infected mice lacking the TNF receptor p55 (TNFRp55(-/-) mice) or both the TNFRp55 and TNFRp75
Y Wannemuehler et al.
Journal of clinical microbiology, 27(12), 2881-2883 (1989-12-01)
A monoclonal antibody to the lipopolysaccharide of Moraxella bovis is described. In an indirect fluorescent-antibody test, this monoclonal antibody reacted with 39 of 39 strains of M. bovis tested and did not react with 26 nonfermenting gram-negative coccobacilli other than
Sophie E Hussey et al.
Bioscience reports, 33(1), 37-47 (2012-10-12)
Emerging evidence suggests that TLR (Toll-like receptor) 4 and downstream pathways [MAPKs (mitogen-activated protein kinases) and NF-κB (nuclear factor κB)] play an important role in the pathogenesis of insulin resistance. LPS (lipopolysaccharide) and saturated NEFA (non-esterified fatty acids) activate TLR4

Articles

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service