Skip to Content
Merck
All Photos(1)

Key Documents

L4020

Sigma-Aldrich

Monoclonal Anti-LEF-1 (β-Catenin binding domain) antibody produced in mouse

clone REMB1, ~1 mg/mL, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
€417.00

€417.00


Please contact Customer Service for Availability


Select a Size

Change View
0.1 MG
€417.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€417.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

REMB1, monoclonal

form

buffered aqueous solution

mol wt

antigen 55-57 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1-5 μg/mL using whole extracts of human Jurkat cells or transiently expressed human LEF-1 in whole cell lysates from Cos-1 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LEF1(51176)

Immunogen

full length recombinant LEF-1.

Physical form

Solution in phosphate buffered saline, containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service