Skip to Content
Merck
All Photos(1)

Key Documents

L3877

Sigma-Aldrich

D-Leu-Tyr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O4
CAS Number:
Molecular Weight:
294.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)C[C@@H](N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C15H22N2O4/c1-9(2)7-12(16)14(19)17-13(15(20)21)8-10-3-5-11(18)6-4-10/h3-6,9,12-13,18H,7-8,16H2,1-2H3,(H,17,19)(H,20,21)/t12-,13+/m1/s1

InChI key

LHSGPCFBGJHPCY-OLZOCXBDSA-N

Biochem/physiol Actions

D-Leucyltyrosine (D-Leu-Tyr) may be used to support the study of the uptake of oligopeptides by various peptide transport mechanisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Daniel et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 9(11), 1112-1117 (1995-08-01)
Based on the involvement of zinc in hydrolysis of peptides, we hypothesized that Zn2+ may also play a role in peptide transport. To investigate this hypothesis, kidney brush border membrane vesicles (BBMV) were incubated for 30 min with different concentrations
J A Krause et al.
Acta crystallographica. Section B, Structural science, 49 ( Pt 1), 123-130 (1993-02-01)
L-Leu-L-Tyr, (I), C15H22N2O4, M(r) = 294.35, crystallizes from MeOH/5% dimethyl sulfoxide in the orthorhombic space group P2(1)2(1)2(1). a = 5.644 (1), b = 12.094 (3), c = 22.548 (4) A, V = 1539.0 (5) A3, Z = 4, Dx =
Pedro Lameiras et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 212(1), 161-168 (2011-08-02)
NMR of weakly polar analytes in an apolar ultraviscous solvent has recently been proposed for mixture analysis as a pertinent alternative to the DOSY experiment. The present article reports the first use of glycerol and glycerol carbonate as polar solvents
Conceição Fonseca et al.
Journal of mass spectrometry : JMS, 44(5), 681-693 (2009-01-07)
The exposure of peptides and proteins to reactive hydroxyl radicals results in covalent modifications of amino acid side-chains and protein backbone. In this study we have investigated the oxidation the isomeric peptides tyrosine-leucine (YL) and leucine-tyrosine (LY), by the hydroxyl
Pedro Domingues et al.
Biomedical chromatography : BMC, 26(1), 51-60 (2011-03-25)
Free radical species are generally short-lived due to their high reactivity and thus direct measurement and identification are often impossible. In this study we used a spin trap, 5,5-dimethyl-1-pyrroline-N-oxide (DMPO), to trap radical intermediates formed during the oxidation of isomeric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service