Skip to Content
Merck
All Photos(1)

Key Documents

L3377

Sigma-Aldrich

Leu-D-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N2O3
CAS Number:
Molecular Weight:
244.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable

storage temp.

−20°C

SMILES string

CC(C)C[C@H](N)C(=O)N[C@H](CC(C)C)C(O)=O

InChI

1S/C12H24N2O3/c1-7(2)5-9(13)11(15)14-10(12(16)17)6-8(3)4/h7-10H,5-6,13H2,1-4H3,(H,14,15)(H,16,17)/t9-,10+/m0/s1

InChI key

LCPYQJIKPJDLLB-VHSXEESVSA-N

Application

Leu-D-Leu may be used as a reference peptide in the development of separation and detection technologies such as CE-ESI-MS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongzhi Ye et al.
Electrophoresis, 31(20), 3400-3406 (2010-09-30)
In this article, an approach has been developed for the analysis of some small peptides with similar pI values by CE-ESI-MS based on the online concentration strategy of dynamic pH junction. The factors affected on the separation, detection and online
Joachim Vater et al.
Rapid communications in mass spectrometry : RCM, 23(10), 1493-1498 (2009-04-08)
An innovative technique to investigate the intermediates involved in the biosynthesis of the lipoheptapeptide surfactin from Bacillus subtilis OKB105 combining whole-cell matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS) with targeted generation of knock-out mutants was demonstrated. This method allows efficient
S N Mitra et al.
Biopolymers, 34(9), 1139-1143 (1994-09-01)
The crystal structure of a dipeptide L-leucyl-L-leucine (C12H24N2O3) has been determined. The crystals are monoclinic, space group P2(1), with a = 5.434(4) A, b = 15.712(7) A, c = 11.275(2) A, beta = 100.41(1) degrees, and Z = 2. The
Pim W J M Frederix et al.
Dalton transactions (Cambridge, England : 2003), 41(42), 13112-13119 (2012-09-20)
A [FeFe]-hydrogenase model compound (µ-S(CH(2))(3)S)Fe(2)(CO)(4)(PMe(3))(2) [1] has been encapsulated in a low molecular weight (LMW) hydrogelator (Fmoc-Leu-Leu). Linear infrared absorption spectroscopy, gel melting and ultrafast time-resolved infrared spectroscopy experiments reveal significant contrasts in chemical environment and photochemistry between the encapsulated
Minghua Lu et al.
Electrophoresis, 30(13), 2273-2279 (2009-07-03)
The objective of this study was to prepare a coated capillary to achieve rapid analysis of peptides and amino acids (AAs) by CE-ESI-MS. The coated capillary was prepared by chemical modification of the fused silica (FS) capillary with 2-acrylamido-2-methyl-1-propanesulfonic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service