Skip to Content
Merck
All Photos(1)

Documents

L3169

Sigma-Aldrich

LP 12 hydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

4-(2-Diphenyl)-N-(1,2,3,4-tetrahydronaphthalen-1-yl)-1- piperazinehexanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H39N3O·HCl · xH2O
Molecular Weight:
518.13 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >5 mg/mL
H2O: 6 mg/mL

storage temp.

−20°C

SMILES string

O.Cl.O=C(CCCCCN1CCN(CC1)c2ccccc2-c3ccccc3)NC4CCCc5ccccc45

InChI

1S/C32H39N3O.ClH.H2O/c36-32(33-30-18-11-15-26-14-6-7-16-28(26)30)20-5-2-10-21-34-22-24-35(25-23-34)31-19-9-8-17-29(31)27-12-3-1-4-13-27;;/h1,3-4,6-9,12-14,16-17,19,30H,2,5,10-11,15,18,20-25H2,(H,33,36);1H;1H2

InChI key

VOKWUIAIZYWMDR-UHFFFAOYSA-N

Biochem/physiol Actions

LP 12 dihydrochloride is a 5-HT7 receptor agonist displaying selectivity over D2, 5-HT1A and 5-HT2A receptors (Ki values are 0.13, 224, 60.9 and 1464 nM, respectively. It also displays selectivity over D2, 1723; over 5-HT1A, 468; over 5-HT2A, 11261; and high potency. Sigma has added LP 44 (L9793) which shows significantly lower D2 selectivity (7.3 nM/0.22 nM for 33-fold), lower selectivity over 5-HT1A, 240; and over 5-HT2A, 1482. LP 12 and LP 44 seem to share a common intermediate.

comparable product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service