Skip to Content
Merck
All Photos(1)

Key Documents

L2781

Sigma-Aldrich

Lead subacetate

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(C2H3O2)2 · 2Pb[OH]2
CAS Number:
Molecular Weight:
807.72
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

composition

Basic lead (PbO), ≥33.0%

impurities

≤0.02% Insoluble in dilute acetic acid
≤1.0% Insoluble in water

loss

≤1.5% loss on drying, 105°

anion traces

chloride (Cl-): ≤0.003%
nitrate and nitrite (as NO3-): passes test (limit about 0.003%)

cation traces

Ca: ≤0.01%
Cu: ≤0.002%
Fe: ≤0.002%
K: ≤0.02%
Na: ≤0.05%

application(s)

food and beverages

SMILES string

O[PbH2]O.O[PbH2]O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.4H2O.3Pb/c2*1-2(3)4;;;;;;;/h2*1H3,(H,3,4);4*1H2;;;/q;;;;;;3*+2/p-6

InChI key

VLOJXAQYHIVPFI-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

For sugar analysis

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service