Skip to Content
Merck
All Photos(1)

Documents

L119

Sigma-Aldrich

L-703,606 oxalate salt hydrate

solid

Synonym(s):

cis-2-(Diphenylmethyl)-N-[(2-iodophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine oxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29IN2 · C2H2O4 · xH2O
CAS Number:
Molecular Weight:
598.47 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

storage condition

desiccated

color

white

solubility

DMSO: >10 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 11 mg/mL
H2O: slightly soluble <2 mg/mL

SMILES string

O.OC(=O)C(O)=O.Ic1ccccc1CN[C@@H]2C3CCN(CC3)[C@@H]2C(c4ccccc4)c5ccccc5

InChI

1S/C27H29IN2.C2H2O4.H2O/c28-24-14-8-7-13-23(24)19-29-26-22-15-17-30(18-16-22)27(26)25(20-9-3-1-4-10-20)21-11-5-2-6-12-21;3-1(4)2(5)6;/h1-14,22,25-27,29H,15-19H2;(H,3,4)(H,5,6);1H2/t26-,27-;;/m1../s1

InChI key

NJAUFRHNYIJKDW-AJUXDCMMSA-N

Gene Information

human ... TACR1(6869)

Related Categories

Biochem/physiol Actions

Potent and selective non-peptide NK-1 tachykinin receptor antagonist.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Cascieri et al.
Molecular pharmacology, 42(3), 458-463 (1992-09-01)
We have synthesized a potent, selective, radioiodinated antagonist of the human neurokinin-1 (NK1) receptor and have characterized its binding to the cloned receptor expressed in Chinese hamster ovary cells. (cis)-2-(Diphenylmethyl)-N-[(2-iodophenyl)-methyl]-1- azabicyclo[2.2.2]octan-3-amine (L-703606) inhibits binding of 125I-Tyr8-substance P to the human
T M Fong et al.
The Journal of biological chemistry, 267(36), 25664-25667 (1992-12-25)
To identify the molecular determinants of ligand-receptor interactions, the extracellular domain of the human neurokinin-1 receptor was systematically substituted with the corresponding sequences from the other two neurokinin receptor subtypes. Three residues within the first extracellular segment and 2 residues
Ki Bum Um et al.
eLife, 10 (2021-08-20)
Midbrain dopamine (DA) neurons are slow pacemakers that maintain extracellular DA levels. During the interspike intervals, subthreshold slow depolarization underlies autonomous pacemaking and determines its rate. However, the ion channels that determine slow depolarization are unknown. Here we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service