Skip to Content
Merck
All Photos(1)

Key Documents

J2455

Sigma-Aldrich

JNK1, active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

JNK, JNK1A1, JNK21B1/2, PRKM8, SAPK1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€494.00

€494.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€494.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€494.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

92-124 nmol/min·mg

mol wt

~71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Mapk8(26419)

Biochem/physiol Actions

As a member of the MAP kinase family, JNK1 is known to play a role in the induction of apoptosis through various stimuli such as TNFa.JNK1 also is influences cell migration via phosphorylation of paxillin.JNK1 is also a crucial mediator of obesity and insulin resistance and a potential target for therapeutics.
JNK1 (c-Jun N-terminal kinase 1) is a member of the MAPK (mitogen activated protein kinase) family that controls gene expression by phosphorylation of transcription factors.[1] It is linked with apoptosis, brain development and learning ability.[2] JNK1 also influences cell migration via phosphorylation of paxillin.[3] JNK1 also regulates obesity and insulin resistance.[4]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prescott T Leach et al.
Neurobiology of learning and memory, 118, 162-166 (2014-12-30)
Increased training often results in stronger memories but the neural changes responsible for these stronger memories are poorly understood. It is proposed here that higher levels of training that result in stronger memories recruit additional cell signaling cascades. This study
Hélène Henrie et al.
The Journal of cell biology, 219(7) (2020-06-04)
The stress-induced c-Jun N-terminal kinase (JNK) controls microtubule dynamics by enhancing both microtubule growth and rescues. Here, we show that upon cell stress, JNK directly phosphorylates the microtubule rescue factor CLIP-170 in its microtubule-binding domain to increase its rescue-promoting activity.
Jiro Hirosumi et al.
Nature, 420(6913), 333-336 (2002-11-26)
Obesity is closely associated with insulin resistance and establishes the leading risk factor for type 2 diabetes mellitus, yet the molecular mechanisms of this association are poorly understood. The c-Jun amino-terminal kinases (JNKs) can interfere with insulin action in cultured
Cai Huang et al.
Nature, 424(6945), 219-223 (2003-07-11)
The c-Jun amino-terminal kinase (JNK) is generally thought to be involved in inflammation, proliferation and apoptosis. Accordingly, its substrates are transcription factors and anti-apoptotic proteins. However, JNK has also been shown to be required for Drosophila dorsal closure, and MAP
C Tournier et al.
Science (New York, N.Y.), 288(5467), 870-874 (2000-05-08)
The c-Jun NH2-terminal kinase (JNK) is activated when cells are exposed to ultraviolet (UV) radiation. However, the functional consequence of JNK activation in UV-irradiated cells has not been established. It is shown here that JNK is required for UV-induced apoptosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service