IST125080SS
QuickSeal Micro™ self adhesive sealing film
LabSheet™, W × L 80 mm × 135 mm, sterile, pack of 100 sheets
Synonym(s):
Self adhesive seal, plate sealing film, low strength plate seal
About This Item
Recommended Products
sterility
sterile
packaging
pack of 100 sheets
manufacturer/tradename
IST Scientific IST-125-080SS
technique(s)
ELISA: suitable
W × L
80 mm × 135 mm
suitability
suitable for ELISA applications
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Sample storage (aqueous samples only)
- Suitable for all plate types
Features and Benefits
- ELISA assays
- Non-pierceable
- Peelable
- Seal integrity range: -20 °C to 80 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service