Skip to Content
Merck
All Photos(1)

Documents

I9385

Sigma-Aldrich

Anti-Human IgM (μ-chain specific) antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:20,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgM is a pentameric immunoglobulin, with each monomer having a light and heavy chain. The two types of IgM comprise natural and adaptive.

Application

Anti-Human IgM (μ-chain specific) antibody produced in rabbit has been used in enzyme linked immunosorbent assay.

Biochem/physiol Actions

IgM antibodies are present as pentamers in the serum and are produced in response to antigens.
IgM serves as prime antibody in providing defense. It activates complement C1q. It is regarded as a marker for identifying tumor-associated antigens. Hyper-IgM syndrome is implicated with high IgM levels accompanied by low IgA, IgG and IgE levels.

Preparation Note

treated to remove lipoproteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human IgM pentamer is a mushroom-shaped molecule with a flexural bias
Czajkowsky DM and Shao Z
Proceedings of the National Academy of Sciences of the USA, 106(35), 14960-14965 (2009)
Natural and adaptive IgM antibodies in the recognition of tumor-associated antigens of breast cancer
Diaz-Zaragoza M, et al.
Oncology Reports, 34(3), 1106-1114 (2015)
Evaluation of a patient with hyper-IgM syndrome
Uygungil B, et al.
The Journal of Allergy and Clinical Immunology, 129(6), 1692-1692 (2012)
Antibodies reacting with human immunoglobulin in sera from autoimmune thyroid disease patients as a risk factor for false positive results in IgA assessment
Haroun M and El-Masry MH
Central European Journal of Immunology, 33, 208-212 (2008)
E J Wiersma et al.
Journal of immunology (Baltimore, Md. : 1950), 160(12), 5979-5989 (1998-06-24)
Previous studies have discerned two forms of polymeric mouse IgM: moderately cytolytic (complement-activating) pentamer, which contains J chain, and highly cytolytic hexamer, which lacks J chain. To investigate the relationships among polymeric structure, J chain content, and cytolytic activity, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service