Skip to Content
Merck
All Photos(1)

Key Documents

I8388

Inoculating Loop

loop size 10 μL

Synonym(s):

inoculating loops, inoculation loop, inoculation loops

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
€434.00

€434.00


Please contact Customer Service for Availability


Select a Size

Change View
500 EA
€434.00

About This Item

UNSPSC Code:
41161502
NACRES:
NB.21

€434.00


Please contact Customer Service for Availability

material

polystyrene

sterility

sterile; γ-irradiated

packaging

pack of 500 ea

manufacturer/tradename

Simport L200-2A

loop size

10 μL

color

color-coded yellow

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Loops are smooth and flexible to facilitate uniform streaking without damaging the gel surface. Disposable inoculating loops do not require flaming and thus eliminate the risk of infection. Loops are not calibrated.

Packaging

In tray packs containing 10 loops

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What are the materials of this item?

    1 answer
    1. As listed in the 'PROPERTIES' section, these inoculating loops are made from polystyrene.

      Helpful?

  2. What are the significance features that distinguish size 10 ul inoculation loop from size 1ul? Thank for you.

    1 answer
    1. The difference between the 10 ul inoculation loop from the 1ul inoculation loop is the loop size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service