Skip to Content
Merck
All Photos(1)

Documents

I4022

Sigma-Aldrich

Hydrogen ionophore II

Synonym(s):

4-Nonadecylpyridine, ETH 1907, Proton ionophore II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H43N
CAS Number:
Molecular Weight:
345.60
Beilstein:
217205
MDL number:
UNSPSC Code:
12171500

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCc1ccncc1

InChI

1S/C24H43N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-24-20-22-25-23-21-24/h20-23H,2-19H2,1H3

InChI key

UMOIOBOWJASTRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service