Skip to Content
Merck
All Photos(1)

Key Documents

I1159

Sigma-Aldrich

trans-4-Iodo-4′-boranyl-chalcone

≥97% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12BIO3
CAS Number:
Molecular Weight:
377.97
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

solubility

DMSO: soluble >30 mg/mL

SMILES string

OB(O)c1ccc(cc1)C(=O)\C=C\c2ccc(I)cc2

Biochem/physiol Actions

MDM2 (mouse double minute 2) inhibitor; immunomodulator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service