Skip to Content
Merck
All Photos(1)

Key Documents

I0779

Sigma-Aldrich

Interleukin-2 Soluble Receptor α human

>97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

CD25, IL-2 sRα, Tac antigen

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€197.00

€197.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 μG
€197.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

€197.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

0.5-1.0 mg per mL ED50

mol wt

36 kDa by SDS-PAGE

packaging

pkg of 5 μg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL2RA(3559)

Biochem/physiol Actions

Interleukin-2 (IL-2), a cytokine, enhances Th (T helper) cell differentiation and effector responses. In addition, it helps in immune tolerance. IL-2 binds to the high affinity receptor, formed of IL2RA (interleukin 2 receptor subunit α), IL2RB (interleukin 2 receptor subunit β) and γ-chain. Polymorphism in IL2RA is associated with multiple sclerosis.[1] Null mutation in IL2RA results in immune dysregulation.[2]

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.25 mg bovine serum albumin.

Analysis Note

The bioactivity is measured by its ability to inhibit the IL-2-dependent proliferation of a human megakaryocytic leukemic cell line, M07e.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Goudy et al.
Clinical immunology (Orlando, Fla.), 146(3), 248-261 (2013-02-19)
Cell-surface CD25 expression is critical for maintaining immune function and homeostasis. As in few reported cases, CD25 deficiency manifests with severe autoimmune enteritis and viral infections. To dissect the underlying immunological mechanisms driving these symptoms, we analyzed the regulatory and
G C Avanzi et al.
British journal of haematology, 69(3), 359-366 (1988-07-01)
A new human leukaemic cell line (M-O7) with the phenotypic characteristics of CFU-mega is described. Its cells are positive for T200 leucocyte common antigen (LCA) and negative with MAbs recognizing T and B cells and mature myelomonocytic antigens. In contrast
Felix J Hartmann et al.
Nature communications, 5, 5056-5056 (2014-10-04)
Genome-wide association studies implicate dysregulation of immune mechanisms in the pathogenesis of multiple sclerosis (MS). Particularly, polymorphisms in genes involved in T helper (TH) cell differentiation are associated with risk of developing MS. However, the underlying mechanism by which these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service