Skip to Content
Merck
All Photos(1)

Key Documents

HUMANHLPB-0002572

Sigma-Aldrich

Frozen Human Normal PBMC

human peripheral blood (mononuclear cells)

Synonym(s):

HLA typed, Immune Cells, PBMC, Peripheral Blood Mononuclear Cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€218.00

€218.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
€218.00

About This Item

UNSPSC Code:
41106514

€218.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Frozen Human Normal PBMC, 20 Million Cells/per vial- HLA Typed

biological source

human peripheral blood (mononuclear cells)

form

frozen liquid

packaging

vial of 20 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

We are pleased to offer these products from BioIVT.

Peripheral Blood Mononuclear Cells (PBMCs) are a subset ofleukocytes (or White Blood Cell, WBC), which are integral components of thebody′s immune system that fight disease. PBMCs consist of a mixture oflymphocytes (T cells, B cells, and NK cells) and monocytes and are isolatedfrom peripheral whole blood via density gradient centrifugation and red bloodcell lysis.

Used for a variety of applications, PBMCs arefrequently utilized in drug discovery/development, assayvalidation/development, and other immunology applications.

Packaging

20 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Functional, viable cryopreserved PBMCs offer a convenient alternative to fresh PBMCs in cell-based assays and immunological studies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service