Skip to Content
Merck
All Photos(2)

Key Documents

HT904

Sigma-Aldrich

Safranin O Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
Product HT904 is not currently sold in your country. Contact Technical Service

form

solution

Quality Level

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

pH

8.5

application(s)

hematology
histology

storage temp.

room temp

Application

Intended for use in Sigma Gram Stain Procedure, HT90.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.0 °F - closed cup

Flash Point(C)

37.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rebecca E Wilusz et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 26(4), 504-512 (2007-12-01)
Damage or loss of the meniscus is associated with progressive osteoarthritic degeneration of the knee joint. Injured and degenerative joints are characterized by elevated levels of the pro-inflammatory cytokine interleukin-1 (IL-1), which with prolonged exposure can induce catabolic and anti-anabolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service