Skip to Content
Merck
All Photos(1)

Key Documents

HT30116

Sigma-Aldrich

Mucicarmine Stock Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
NACRES:
NA.47
Product HT30116 is not currently sold in your country. Contact Technical Service

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Stains mucin of epithelial origin in tissue.

Other Notes

Carmine, certified, 10 g/L, aluminum hydroxide, 10g/L, and aluminum chloride, 5g/L, in ethanol, 50% w/v. Southgates′s modification of Mayer′s mucicarmine procedure.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service