Skip to Content
Merck
All Photos(2)

Key Documents

HT1102128

Sigma-Aldrich

Eosin Y Solution, Aqueous

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
€320.00

€320.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 L
€320.00

About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

€320.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.5 % (w/v) in water

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Other Notes

Certified Eosin Y, 0.5% (w/v) in water. Not acidified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service