Skip to Content
Merck
All Photos(2)

Key Documents

HS120651

Reagent Reservoir

capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)

Sign Into View Organizational & Contract Pricing

Select a Size

200 EACH
€176.00

€176.00


Please contact Customer Service for Availability


Select a Size

Change View
200 EACH
€176.00

About This Item

UNSPSC Code:
41121522
NACRES:
NB.01

€176.00


Please contact Customer Service for Availability

material

V-bottom bottom
polystyrene

sterility

sterile; electron beam irradiated

feature

Divided as 10 and 15 mL

packaging

pack of 200 (packed as 5 ea)

manufacturer/tradename

Heathrow Scientific 120651

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.

Features and Benefits

  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service