Skip to Content
Merck
All Photos(1)

Documents

HS120480

Universal carousel pipette stand

blue to green, 8 of each color for top + 8 blue bottoms

Synonym(s):

pipet stand, pipette carousel, pipettor stand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122801
NACRES:
NB.51

feature

Holds 16 x pipettors

manufacturer/tradename

Heathrow Scientific HS120480

color

blue to green, 8 of each color for top + 8 blue bottoms

Looking for similar products? Visit Product Comparison Guide

General description

Pipettes never looked so good, on your bench top

Hold and store pipettes in style with the new universal carousel pipette stand that holds up to 8 single channel, 4 multi channel, or a combination of both pipettes. A sleek and modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace making it truly ‘universal′ while adding a pop of color to a bench top. Inserts on the upper holders can be added or removed to accommodate the fit of different of pipette widths. One handed removal or replacement of pipettes is made easy with a rotating head and stable weighted base. Moving the stand while keeping pipettes in place is simple with a quick grip handle allowing a user to grab and go. The pipette stands come with additional upper leaves to enable multiple color coding configuration options as well as a change in color to your work space, for a bit more fun.

  • Ideal for labs that do not wish to be tied into a specific brand of pipette, or already have multiple brands in use
  • Weighted base allows for easy removal or replacement of pipettes, with one hand
  • Pipettes are held in a vertical position to help prevent contamination
  • Conveniently rotate stand head to select or store pipettes
  • Quick grip handle to move pipette stand (loaded or unloaded)
  • Assorted color options allow for color coding pipette location: Blue/Green; Purple/Clear; Assorted/Clear
  • The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
  • Made of ABS (Acrylonitrile Butadiene Styrene)
  • US Patent Pending


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service