Skip to Content
Merck
All Photos(1)

Key Documents

HS120261

Blood Collection Tray with rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€100.00

€100.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€100.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

€100.00


Please contact Customer Service for Availability

material

acrylonitrile-butadiene-styrene (ABS)

feature

holds Phlebotomy, I.V. start or catheter insertion supplies

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific HS120261

General description

The Blood Collection Tray is lightweight, heavy on organization. Organize your phlebotomy, I.V. start or catheter insertion supplies with this spacious, robust and easy to clean carry tray.

Features and Benefits

  • Customize the compartments to suit your personal needs with the dividers provided
  • Comfort grip handle tested to hold up to 10 kg
  • Lightweight easy-to-clean ABS construction
  • Includes a 17 mm well, 60-place tube rack

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service