Skip to Content
Merck
All Photos(1)

Key Documents

HS120073

TrueNorth® Mini Cooler

for tubes, 0.5 ‑ 2 mL, with clear lid, temp. limit -20 °C, size 12 wells, blue polycarbonate

Synonym(s):

cooling tube rack, tube chiller

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€145.00

€145.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€145.00

About This Item

UNSPSC Code:
41103025
NACRES:
NB.22

€145.00


Please contact Customer Service for Availability

material

blue polycarbonate

feature

for tubes 0.5 ‑ 2 mL

parameter

-20 °C temp. limit

L × W × H

145 × 105 × 100 mm

size

12 wells

Looking for similar products? Visit Product Comparison Guide

General description

Protect temperature sensitive samples at the bench or in the freezer. Mini Coolers are ideal for keeping PCR reagents, enzymes, and other biochemical reagents cool while working at the bench. Filled with insulating gel, the mini coolers can also be used for ccontrolled sample freezing and for longer temperature retention samples.

Features and Benefits

  • polycarbonate construction with rubber non-slip feet
  • gridded and keyed one-position lids for easy sample identification
  • 0.5 mL tube adapters supplied as standard with the 0.5-2.0 mL tube size versions
  • temperature retention graph supplied with each of the 7 styles
  • versions available or 0.5 and 1.5/2.0 mL microcentrifuge tubes and cyrogenic vials and a 96-place version for 0.2 mL tubes and assay plates

Legal Information

TrueNorth is a registered trademark of Heathrow Scientific LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service