Skip to Content
Merck
All Photos(1)

Key Documents

HPA052258

Sigma-Aldrich

Anti-IL17A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CTLA8, Anti-IL-17, Anti-IL-17A, Anti-IL17, Anti-interleukin 17A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

NPGCPNSEDKNFPRTVMVNLNIHNRNTNTNPKRSSDYYNRSTSPWNLHRNEDPERYPSVIWEAKCRHLGCIN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL17A(3605)

General description

The gene interleukin 17A (IL17A), also referred to as IL-17, is mapped to human chromosome 6p12.2. It contains three exons. IL17A is a proinflammatory cytokine produced by activated T cells.[1]

Immunogen

interleukin 17A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry[2] against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Interleukin 17A (IL17A) activates proinflammatory cytokines, chemokines and cell adhesion molecules in several cell types such as, macrophages, dendritic cells, T cells, synovial cells and endothelial cells. Overexpression of this protein has been associated with various pathologies like rheumatoid arthritis, systemic lupus erythematosus, Behcet′s disease, allograft rejection, nephritic syndrome, asthma and multiple sclerosis. In mice, IL17A is implicated in the development of allergic and autoimmune diseases. Polymorphism in the gene has been associated with lung cancer.[1][3]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78069.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-17 plays an important role in the development of experimental autoimmune encephalomyelitis.
Komiyama Y
Journal of Immunology (2006)
Marlena Janiczek et al.
Journal of immunology research, 2020, 4910595-4910595 (2020-06-17)
Prostate cancer (PCa) is the second most commonly diagnosed malignant tumor and the fifth leading cause of cancer death in men in the world. The most common types of tumors are adenocarcinomas. Prostate cancer is a slow-growing cancer. The incidence
Interleukin-17A and -17F genes polymorphisms in lung cancer.
Kaabachi W
Cytokine (2014)
Fanhang Meng et al.
Inflammation, 37(5), 1799-1805 (2014-05-03)
Myeloid-derived suppressor cells (MDSCs) are negative regulators of the immune response and are in part responsible for the inhibition of the T cell-mediated immune response. A recent paper indicated that MDSCs were involved in prolonged allograft survival in animal models
Qing Li et al.
Cell biochemistry and biophysics, 70(2), 1489-1498 (2014-05-07)
The imbalance of CD4(+)CD25(+) regulatory T (Treg) cells and Th17 cells has shown to be involved in pathogenesis of atherosclerosis and acute coronary syndrome [ACS, including unstable angina (UA) and acute myocardial infarction (AMI)]. The purpose of this study is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service