Skip to Content
Merck
All Photos(5)

Key Documents

HPA044948

Sigma-Aldrich

Anti-MIIP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Flj12438, Anti-Iip45, Anti-Migration and invasion inhibitory protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SDTLALPRHCLLGWDIFPPKSEKSSAPRNLDLWSSVSAEAQHQKLSGTSSPFHPASPMQMLPPTPTWSVPQVPRPHVPRQKP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MIIP(60672)

General description

Migration and invasion inhibitory protein (MIIP), also called insulin-like growth factor binding protein 2 (IGFBP2), is expressed in fetal tissues and astroglial cells. It has an insulin-like growth factor (IGF) binding motif. It is a tumor-suppressor gene and is mapped to human chromosome 1p36.22. It has a C-terminal polyproline domain.

Immunogen

migration and invasion inhibitory protein recombinant protein epitope signature tag (PrEST)

Application

Anti-MIIP antibody produced in rabbit has been used in western blotting and in immunoprecipitation detection of MIIP in colorectal cancer cells.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Migration and invasion inhibitory protein (MIIP), binds to insulin-like growth factor (IGF) and modulates cancer progression. It is under expressed in brain tumor, invasive glioblastoma multiforme. MIIP is a tumor suppressor gene and inhibits rat sarcoma (Ras)-related C3 botulinum toxin substrate 1- guanosine triphosphate (Rac1-GTP) mediated cell migration in endometrial carcinoma. Single nucleotide polymorphism in MIIP is associated with increased risk of breast cancer. MIIP inhibits non-small cell lung cancer progression by lowering epidermal growth factor receptor (EGFR) expression. Haploinsufficiency of MIIP is implicated in the colorectal tumor progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76605

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Definition of a functional single nucleotide polymorphism in the cell migration inhibitory gene MIIP that affects the risk of breast cancer
Song F, et al.
Cancer Research, 70(3), 1024-1032 (2010)
PRP4 kinase induces actin rearrangement and epithelial-mesenchymal transition through modulation of the actin-binding protein cofilin
Islam SU, et al.
Experimental Cell Research, 369(1), 158-165 (2018)
IIp45, an insulin-like growth factor binding protein 2 (IGFBP-2) binding protein, antagonizes IGFBP-2 stimulation of glioma cell invasion
Song SW, et al.
Proceedings of the National Academy of Sciences of the USA, 100(24), 13970-13975 (2003)
MIIP remodels Rac1-mediated cytoskeleton structure in suppression of endometrial cancer metastasis
Wang Y, et al.
Journal of Hematology & Oncology, 9(1), 112-112 (2016)
Yan Sun et al.
The Journal of pathology, 241(1), 67-79 (2016-10-16)
The gene encoding migration and invasion inhibitory protein (MIIP), located on 1p36.22, is a potential tumour suppressor gene in glioma. In this study, we aimed to explore the role and mechanism of action of MIIP in colorectal cancer (CRC). MIIP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service