Skip to Content
Merck
All Photos(4)

Key Documents

HPA040967

Sigma-Aldrich

Anti-DNAJB9 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dnaj (hsp40) homolog, subfamily b, member 9, Anti-Mdg1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

GGSSRQRHHFQEFSFGGGLFDDMFEDMEKMFSFSGFDSTNQHTVQTENRFHGSSKHCRTVTQRRGNMVTTYTDCSGQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAJB9(4189)

General description

DnaJ homolog subfamily B member 9 (DNAJB9) belongs to the molecular chaperone gene family. It is also termed as Mdg-1 or ERdj4. This protein is located in the endoplasmic reticulum (ER). DNAJB9 is located on human chromosome 7q31.

Immunogen

DnaJ (Hsp40) homolog, subfamily B, member 9 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-DNAJB9 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

DnaJ homolog subfamily B member 9 (DNAJB9) act as an inducible gene at the time of endoplasmic reticulum (ER) stress. J domain of DNAJB9 is required for the interaction with Hsp70s. It is considered as a specific immunohistochemical marker for fibrillary glomerulonephritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81907

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samih H Nasr et al.
Kidney international reports, 3(1), 56-64 (2018-01-18)
Fibrillary glomerulonephritis (FGN) is a rare disease with unknown pathogenesis and a poor prognosis. Until now, the diagnosis of this disease has required demonstration of glomerular deposition of randomly oriented fibrils by electron microscopy that are Congo red negative and
Surendra Dasari et al.
Journal of the American Society of Nephrology : JASN, 29(1), 51-56 (2017-11-04)
Fibrillary GN (FGN) is a rare primary glomerular disease. Histologic and histochemical features of FGN overlap with those of other glomerular diseases, and no unique histologic biomarkers for diagnosing FGN have been identified. We analyzed the proteomic content of glomeruli
DNAJB9 is a specific immunohistochemical marker for fibrillary glomerulonephritis.
Nasr SH, et al.
Kidney international reports, 3(1), 56-64 (2018)
MDG1/ERdj4, an ER?resident DnaJ family member, suppresses cell death induced by ER stress.
Kurisu J, et al.
Genes Cells, 8(2), 189-202 (2003)
Mireille El Ters et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 76(4), 500-510 (2020-05-18)
Fibrillary glomerulonephritis (FGN) is a rare glomerular disease that often progresses to kidney failure requiring kidney replacement therapy. We have recently identified a novel biomarker of FGN, DnaJ homolog subfamily B member 9 (DNAJB9). In this study, we used sequential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service