Skip to Content
Merck
All Photos(4)

Key Documents

HPA040505

Sigma-Aldrich

Anti-FERMT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FERMT2, Anti-Fermitin family homolog 2 (Drosophila), Anti-KIND2, Anti-Mig-2, Anti-PLEKHC1, Anti-UNC112B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MADSSYNLEVQNILSFLKMQHLNPDPQLIPEQITTDITPECLVSPRYLKKYKNKQITARILEA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLEKHC1(10979)

General description

Fermitin family member 2 (FERMT2), also known as kindlin-2 or pleckstrin homology domain-containing family C member 1 (PLEKHC1), is encoded by the gene mapped to human chromosome 14q22.1. FERMT2 is a member of the kindlin protein family and has a ubiquitous expression pattern.

Immunogen

fermitin family homolog 2 (Drosophila) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fermitin family member 2 (FERMT2) acts as a focal adhesion protein and controls breast cancer progression by promoting genome instability. FERMT2, as an integrin-interacting protein, plays a vital role in integrin activation and integrin-mediated cell-extracellular matrix (ECM) interaction. FERMT2 expression is associated with poor prognosis of patients with hepatocellular carcinoma (HCC) and pancreatic ductal adenocarcinomas (PDACs). Therefore, FERMT2 can be considered as a potential biomarker for patients with HCC and PDACs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81813

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu-Yue Zhang et al.
Oncotarget, 7(26), 39044-39050 (2016-10-27)
A recent meta-analysis of genome-wide association studies (GWAS) in population of Caucasian identified a single nucleotide polymorphism (SNP) rs17125944 in the FERMT2 gene as a new susceptibility locus for late-onset Alzheimer's disease (LOAD). In order to validate the association of
Yong-Sheng Ge et al.
Pathology, research and practice, 211(3), 198-202 (2015-01-27)
The association of aberrant expression of Kindlin-2 with tumor progression has been reported in recent years. The purpose of this study was to investigate the expression of Kindlin-2 in hepatocellular carcinoma (HCC), and to evaluate its clinical and prognostic significance.
Prawej Mahawithitwong et al.
Pancreas, 42(4), 663-669 (2013-03-20)
Kindlin-2 is a novel focal adhesion protein reported to be expressed in breast, lung, and gastric cancers. This study aimed to investigate the significance of kindlin-2 expression in pancreatic ductal adenocarcinomas (PDACs). We performed immunohistochemical analysis on kindlin-2 on PDAC
Ting Zhao et al.
Cancer letters, 330(2), 208-216 (2012-12-06)
Kindlin-2, as a focal adhesion protein, has been found to regulate tumor progression. However, the mechanism underlying Kindlin-2 regulation of tumor progression is largely unknown. Here, we report that Kindlin-2 regulates breast cancer cell proliferation, apoptosis and chromosomal abnormalities in
Chiea Chuen Khor et al.
Nature genetics, 48(5), 556-562 (2016-04-12)
Primary angle closure glaucoma (PACG) is a major cause of blindness worldwide. We conducted a genome-wide association study (GWAS) followed by replication in a combined total of 10,503 PACG cases and 29,567 controls drawn from 24 countries across Asia, Australia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service