Skip to Content
Merck
All Photos(7)

Key Documents

HPA035787

Sigma-Aldrich

Anti-TMPRSS2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PRSS10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GSPPAIGPYYENHGYQPENPYPAQPTVVPTVYEVHPAQYYPSPVPQYAPRVLTQASNPVVCTQPKSPSGTVCTSKT

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMPRSS2(7113)

General description

Transmembrane serine protease 2 (TMPRSS2) belongs to the membrane-anchored serine proteases (MASP) family, which is mainly expressed in the prostate. The gene TMPRSS2 codes for a prostate-specific androgen-responsive protease. This multimeric protein TMPRSS2 consists of a serine protease domain, a scavenger receptor cysteine-rich (SRCR) domain, a low-density lipoprotein receptor class A (LDLRA) domain and a transmembrane domain. The TMPRSS2 gene is located on human chromosome 21q22.3.

Immunogen

transmembrane protease, serine 2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-TMPRSS2 antibody produced in rabbit has been used for immunohistochemistry at a dilution of 1:200-1:500. It has also been used in western blotting at a working concentration of 0.04-0.4 μg/ml.

Biochem/physiol Actions

Transmembrane serine protease 2 (TMPRSS2) expression level is related to prostate cancer progression. This protein stimulates diverse coronaviruses (CoVs), human metapneumovirus, parainfluenza virus and hepatitis C virus in cell culture. It might lead to a viral spread in the host. TMPRSS2-specific inhibitors exhibit a broad antiviral effect. TMPRSS2 cleaves and stimulates the influenza virus hemagglutinin (HA). It is employed for S protein priming and is known to inhibit the host cell entry of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST87138

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawel Zmora et al.
PloS one, 10(9), e0138380-e0138380 (2015-09-18)
The cellular protease TMPRSS2 cleaves and activates the influenza virus hemagglutinin (HA) and TMPRSS2 expression is essential for viral spread and pathogenesis in mice. Moreover, severe acute respiratory syndrome coronavirus (SARS-CoV) and other respiratory viruses are activated by TMPRSS2. However
Sarah Ibrahim et al.
Islets, 13(3-4), 66-79 (2021-05-11)
The link between COVID-19 infection and diabetes has been explored in several studies since the start of the pandemic, with associations between comorbid diabetes and poorer prognosis in patients infected with the virus and reports of diabetic ketoacidosis occurring with
Sarah M Roelle et al.
PLoS biology, 20(7), e3001738-e3001738 (2022-07-28)
Viral spillover from animal reservoirs can trigger public health crises and cripple the world economy. Knowing which viruses are primed for zoonotic transmission can focus surveillance efforts and mitigation strategies for future pandemics. Successful engagement of receptor protein orthologs is
Samsul Arefin et al.
European journal of clinical investigation, 52(8), e13786-e13786 (2022-04-03)
Individuals with chronic kidney disease are affected by acute respiratory syndrome coronavirus 2 (SARS-CoV-2) due to multiple comorbidities and altered immune system. The first step of the infection process is the binding of SARS-CoV-2 with angiotensin-converting enzyme 2 (ACE2) receptor
A Paoloni-Giacobino et al.
Genomics, 44(3), 309-320 (1997-11-05)
To contribute to the development of the transcription map of human chromosome 21 (HC21), we have used exon trapping from pools of HC21-specific cosmids. Using selected trapped exons, we have identified a novel gene (named TMPRSS2) that encodes a multimeric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service