Skip to Content
Merck
All Photos(9)

Key Documents

HPA029103

Sigma-Aldrich

Anti-RGN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-RC, Anti-SMP30, Anti-regucalcin (senescence marker protein-30)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DAEGKLWVACYNGGRVIRLDPVTGKRLQTVKLPVDKTTSCCFGGKNYSEMYVTCARDGMDPEGLLRQPEAGGIFKITGLGVKGIAPYSY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGN(9104)

General description

Regucalcin protein 30 (SMP-30), is a calcium binding protein. RGN is mapped to human chromosome X and has seven exons. It has three alternatively spliced transcripts corresponding to 20 kDa and 25 kDa proteins. RGN structure shows a six-bladed β-propeller fold. The active site for calcium binding is located in the central tunnel region. It lacks EF-hand calcium binding motif. (RGN), also called senescence marker.

Immunogen

regucalcin (senescence marker protein-30) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Regucalcin (RGN) regulates calcium-mediated signalling events. RGN has gluconolactonase functionality, catalysing L-ascorbic acid (AA) biosynthesis. It recognizes and binds to L-gluconate and mediates lactone-ring cleavage reactions. RGN acts as tumor suppressor and promotes cell cycle arrest in human pancreatic cancer. RGN is less expressed in hepatocellular carcinoma and may play a key role in cytoskeletal organisation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78509

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayoshi Yamaguchi et al.
Translational oncology, 14(1), 100955-100955 (2020-11-25)
Prostate cancer, which is a bone metastatic cancer, is the second leading cause of cancer-related death in men. There is no effective treatment for metastatic prostate cancer. Regucalcin has been shown to contribute as a suppressor in various types of
Regucalcin plays a role in the cytoskeleton regulation of HepG2 cells
Chen J, et al.
Acta biochimica et biophysica Sinica, 49(2), 193-195 (2016)
Masayoshi Yamaguchi et al.
Oncology, 100(7), 399-412 (2022-03-28)
Regucalcin plays a multifunctional role in the regulation of cellular function including metabolism, signaling process, and transcriptional activity in maintaining cell homeostasis. Downregulated expression or activity of regucalcin contributes to the development of malignancies in various types of human cancer.
Crystal structure of human senescence marker protein 30: insights linking structural, enzymatic, and physiological functions
Chakraborti S and Bahnson BJ
Biochemistry, 49(16), 3436-3444 (2010)
Alternatively spliced variants of the regucalcin gene in various human normal and tumor tissues
Murata T and Yamaguchi M
International Journal of Molecular Medicine, 34(4), 1141-1146 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service