Skip to Content
Merck
All Photos(3)

Documents

HPA028997

Sigma-Aldrich

Anti-SLC2A2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GLUT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

PETKGKSFEEIAAEFQKKSGSAHRPKAAVEMKFLGATET

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A2(6514)

General description

Solute carrier family 2 member 2 (SLC2A2) gene codes for glucose transporter 2 (GLUT2). GLUT2 protein is expressed in pancreatic β cells, liver, kidney, intestine, and central nervous system. SLC2A2 gene is located on human chromosome 3q26.2.

Immunogen

solute carrier family 2 (facilitated glucose transporter), member 2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SLC2A2 antibody produced in rabbit has been used in immunofluorescence (1:300) and immunohistochemistry (1:20).

Biochem/physiol Actions

The glucose transporter 2 (GLUT2) protein serves as a low-affinity facilitative glucose transporter. It may play a role in the development of the pancreas. Mutations in the SLC2A2 gene result in permanent neonatal diabetes (PNDM) and Fanconi–Bickel syndrome (FBS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78554

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed K Elsayed et al.
Stem cell research, 54, 102433-102433 (2021-06-26)
Recessive mutations in the glucose transporter gene SLC2A2 (GLUT2) lead to permanent neonatal diabetes (PNDM) and Fanconi Bickel Syndrome (FBS). Here, we generated an induced pluripotent stem cell (iPSC) line, QBRIi012-A, from a 24-month-old boy with FBS and PNDM due
Ahmed K Elsayed et al.
Stem cell research, 44, 101736-101736 (2020-03-09)
Fanconi Bickel Syndrome (FBS) is an autosomal recessive disease resulting from mutations in the SLC2A2 gene, encoding the GLUT2. FBS patients develop diabetes mellitus. Using non-integrating Sendai virus, we generated an induced pluripotent stem cell (iPSC) line, QBRIi007-A, carrying the
Chantal Florence Meier et al.
Journal of pediatric gastroenterology and nutrition, 69(5), 611-618 (2019-07-02)
In newborn rodents, intestinal maturation involves delayed fructose transporter GLUT5 expression until weaning. In jejunoileal atresia (JIA), distal intestinal segments lack exposure to amniotic fluid-containing carbohydrates. We assessed in human newborns, the impact of intestinal maturation and obstruction on mucosal
Richard Jennemann et al.
Glycobiology, 30(9), 722-734 (2020-03-10)
In pancreatic beta cells, the entry of glucose and downstream signaling for insulin release is regulated by the glucose transporter 2 (Glut2) in rodents. Dysfunction of the insulin-signaling cascade may lead to diabetes mellitus. Gangliosides, sialic acid-containing glycosphingolipids (GSLs), have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service