Skip to Content
Merck
All Photos(2)

Key Documents

HPA027755

Sigma-Aldrich

Anti-MAP2K5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-HsT17454, Anti-MAPKK5, Anti-MEK5, Anti-PRKMK5, Anti-mitogen-activated protein kinase kinase 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

IFPRACKPPGERNIHGLKVNTRAGPSQHSSPAVSDSLPSNSLKKSSAELKKILANGQMNEQDIRYRDTLGHGNG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP2K5(5607)

General description

Mitogen-activated protein kinase kinase 5 (MAP2K5) is part of the mitogen-activated protein kinase family. The MAP2K5 gene encodes three protein-coding transcripts, consists of 20 exons and is localized on human chromosome 15q23. MEK5α is expressed in the liver and MEK5β is expressed in terminally differentiated tissues.

Immunogen

mitogen-activated protein kinase kinase 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mitogen-activated protein kinase kinase 5 (MAP2K5) has a role in signaling pathways which are mediated by proteins like brain derived neurotrophic factor (BDNF), insulin-like growth factor 2 (IGF2) and nerve growth factor (NGF). It also responds to various stress stimuli. MEK5α, one of the three MAP2K5 isoforms, activates extracellular signal regulated kinase 5 (ERK5). On the other hand, MEK5β suppresses ERK5 signaling. Upregulation of MEK5α has been linked to tumor growth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77993

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitogen/Extracellular Signal-Regulated Kinase Kinase-5 Promoter Region Polymorphisms Affect the Risk of Sporadic Colorectal Cancer in a Southern Chinese Population
Dechang Diao
Dna and Cell Biology (2012)
The MAP2K5-linked SNP rs2241423 is associated with BMI and obesity in two cohorts of Swedish and Greek children
Mathias Rask-Andersen
BMC Medical Genetics (2012)
Array CGH identifies distinct DNA copy number profiles
of oncogenes and tumor suppressor genes in chromosomaland
microsatellite-unstable sporadic colorectal carcinomas
Silke Lassmann
Journal of Molecular Medicine (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service