Skip to Content
Merck
All Photos(4)

Key Documents

HPA023230

Sigma-Aldrich

Anti-VBP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HIBBJ46, Anti-Prefoldin subunit 3, Anti-VBP-1, Anti-VHL-binding protein 1, Anti-Von Hippel-Lindau-binding protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

DVDSFMKQPGNETADTVLKKLDEQYQKYKFMELNLAQKKRRLKGQIPEIKQTLEILKYMQKKKESTNSMETRFLLADNLYCKASVPPTDKVCLWLGANVMLEYDIDEAQALLEKNLSTATKNLDSLEEDLDFLRDQFTTTEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VBP1(7411)

General description

The gene VBP1 (von Hippel-Lindau binding protein 1) is mapped to human chromsome Xq28. The protein localizes in the nucleus and cytoplasm.

Immunogen

Prefoldin subunit 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VBP1 (von Hippel-Lindau binding protein 1) interacts with the von Hippel-Lindau (VHL) ubiquitin E3 ligase and forms a VHL-elongin B-elongin C complex (VBC). It is associated with the assembly of α- and γ-tubulins. It is shown to connect HIV (human immunodeficiency virus)-1 integrase and VHL, leading to integrase degradation. VBP1 is also a component of the prefoldin chaperone.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74397

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hemberger et al.
Human molecular genetics, 8(2), 229-236 (1999-02-05)
The von Hippel-Lindau (VHL) tumour suppressorgene product is believed to be involved in the down-regulation of transcriptional elongation by preventing the association of elongin B and C with the catalytic subunit elongin A. Alterations in the human VHL gene lead
Aurélie Mousnier et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(34), 13615-13620 (2007-08-19)
HIV-1 integrase, the viral enzyme responsible for provirus integration into the host genome, can be actively degraded by the ubiquitin-proteasome pathway. Here, we identify von Hippel-Lindau binding protein 1(VBP1), a subunit of the prefoldin chaperone, as an integrase cellular binding
H Tsuchiya et al.
Cancer research, 56(13), 2881-2885 (1996-07-01)
The tumor suppressor VHL gene product (pVHL), recently reported to bind to elongins B and C, is thought to regulate transcription elongation. To establish whether the VHL gene may have other functions, we here searched for additional cellular protein(s) that
David Schörghofer et al.
British journal of cancer, 131(3), 468-480 (2024-06-21)
Despite ongoing research and recent advances in therapy, metastatic melanoma remains one of the cancers with the worst prognosis. Here we studied the postsynaptic cell adhesion molecule Neuroligin 4X (NLGN4X) and investigated its role in melanoma progression. We analysed histologic
Yang Xu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(12), 4799-4810 (2013-08-22)
Ubiquitination is an important mechanism for the regulation of diverse cellular functions, including proteolysis and DNA repair. The human MutS family protein hMSH4 functions in meiotic recombinational DNA double-strand break (DSB) repair. It was previously observed that hMSH4 interacts with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service