Skip to Content
Merck
All Photos(6)

Key Documents

HPA022924

Sigma-Aldrich

Anti-LLGL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-DLG4, Anti-Hugl-1, Anti-Human homolog to the D-lgl gene protein, Anti-LLGL, Anti-Lethal recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSPMSIDSATSADTTLDTTGDVTVEDVKDFLGSSEESEKNLRNLAEDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LLGL1(3996)

Immunogen

Lethal recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LLGL1 (lethal(2) giant larvae protein homolog 1) is downregulated in breast, lung, prostate, ovarian cancers and melanomas. Low expression of LLGL1 is linked with lymph node metastasis and advanced stage cancer. In mouse, absence of LLGL1 results in the formation of neuroectodermal tumor in the brain. The gene is mapped to human chromosome 17.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75620

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atlantis Russ et al.
PloS one, 7(10), e47734-e47734 (2012-10-31)
Loss of epithelial polarity is described as a hallmark of epithelial cancer. To determine the role of Hugl1 and Hugl2 expression in the breast, we investigated their localization in human mammary duct tissue and the effects of expression modulation in
Xuefeng Lu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 15(10), 3287-3296 (2009-05-19)
Lethal giant larvae functions as a cell polarity regulator and a tumor suppressor in Drosophila. Its evolutionary conservation implies a tumor suppressor role for its human homologue, Hugl-1. The aims of this study were to characterize Hugl-1 and to determine
Stefan Biesterfeld et al.
Anticancer research, 32(8), 3153-3159 (2012-07-31)
The expression of the human homologue of Drosophila tumour suppressor gene lgl (HUGL-1) in pancreatic cancer was retrospectively assessed in 97 patients with surgically treated pancreatic cancer in order to correlate the HUGL-1 profile with patients' survival. Immunohistochemistry was performed
Kazunori Sasaki et al.
Cell reports, 31(1), 107407-107407 (2020-04-09)
Epithelial cells establish apicobasal polarity by forming tight junctions (TJs) at the apical-lateral boundary, which play fundamental roles in physiological functions. An evolutionarily conserved atypical protein kinase C (aPKC)-partitioning defective (PAR) complex functions as a platform for TJ assembly during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service