Skip to Content
Merck
All Photos(4)

Key Documents

HPA021989

Sigma-Aldrich

Anti-ANAPC11 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APC11, Anti-Anaphase-promoting complex subunit 11, Anti-Cyclosome subunit 11, Anti-Hepatocellular carcinoma-associated RING finger protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SPWCLDHSCDLFGITDQVSADGPRACRQGARRRLPAGVGPVLPLLPHALHPQVAARTAGAAALPHVPPGMEVQGV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANAPC11(51529)

General description

ANAPC11 (Anaphase promoting complex subunit 11) is a E3 ubiquitin ligase consisting of a RING-H2-finger domain with one histidine and seven cysteine residues that coordinate two Zn(2+) ions. It is mapped on the human chromosome 17q25 and is expressed in the cytoplasm and nucleus with discrete accumulation in granular structures in all the cell lines (AML 12, HepG2, and C2C12) transfected.

Immunogen

Anaphase-promoting complex subunit 11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ANAPC11 (Anaphase promoting complex subunit 11) plays a key role in cell division by accelerating ubiquitination of cell-cycle regulators such as cyclin B and securin. It acts as the catalytic subunit of anaphase-promoting complex (APC) and helps to translocate ubiquitin from a ubiquitin-conjugating enzyme (E2) to the substrate. It controls the cell cycle progression through G1 phase of the cell cycle during protein modification.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75676

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A H Chan et al.
Journal of cellular biochemistry, 83(2), 249-258 (2001-09-27)
Yeast Apc11p together with Rbx1 and Roc2/SAG define a new class of RING-H2 fingers in a superfamily of E3 ubiquitin ligases. The human homolog of Apc11p, ANAPC11 was identified during a large-scale partial sequencing of a human liver cancer cDNA
Tong-Shin Chang et al.
Free radical biology & medicine, 37(4), 521-530 (2004-07-17)
The anaphase-promoting complex (APC) is a ubiquitin-protein ligase (E3) that targets cell cycle regulators such as cyclin B and securin for degradation. The APC11 subunit functions as the catalytic core of this complex and mediates the transfer of ubiquitin from
Y-J Shi et al.
Genetics and molecular research : GMR, 11(3), 2814-2822 (2012-09-26)
Anaphase-promoting complex/cyclosome (APC/C) is a key E3 ubiquitin ligase in cell division, which catalyses ubiquitination of cell-cycle regulators. Studying this complex could reveal important information regarding its application in cancer research and therapy. In this study, 4 synthesized small interfering

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service