Skip to Content
Merck
All Photos(2)

Key Documents

HPA021868

Sigma-Aldrich

Anti-ROR2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neurotrophic tyrosine kinase, receptor-related 2, Anti-Tyrosine-protein kinase transmembrane receptor ROR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

FPQPQFIPMKGQIRPMVPPPQLYVPVNGYQPVPAYGAYLPNFYPVQIPMQMAPQQVPPQMVPKPSSHHSGSGSTSTGYVTTAPSNTSMADRAALLSEGADDTQNAPEDGAQSTVQEAEEEEEGSVPETELLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ROR2(4920)

Immunogen

Tyrosine-protein kinase transmembrane receptor ROR2 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ROR2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71452.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ripla Arora et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 243(8), 1037-1045 (2014-04-23)
The noncanonical Wnt receptor and tyrosine kinase Ror2 has been associated with recessive Robinow syndrome (RRS) and dominant brachydactyly type B1. The phenotypes of mouse mutants implicate Ror2 in the development of the heart, lungs, bone, and craniofacial structures, which
Erratum to: Validation of specificity of antibodies for immunohistochemistry: the case of ROR2.
Sean S Q Ma et al.
Virchows Archiv : an international journal of pathology, 469(6), 717-717 (2016-11-01)
Lei Wang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(5), 6085-6090 (2015-11-27)
Primary thyroid lymphoma (PTL) is a rare malignant thyroid tumor; its pathogenesis is closely related to chronic lymphocytic thyroiditis. The different pathological subtypes and stages of PTL have distinct clinical characteristics and prognosis, but the specific reasons are not clear.
Claire Henry et al.
Oncotarget, 6(37), 40310-40326 (2015-10-31)
In recent years, the Wnt signalling pathway has been implicated in epithelial ovarian cancer and its members have potential as diagnostic, prognostic and therapeutic targets. Here we investigated the role of two Wnt receptor tyrosine kinases (RTKs), ROR1 and ROR2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service