Skip to Content
Merck
All Photos(4)

Key Documents

HPA021466

Sigma-Aldrich

Anti-SMAP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Stromal membrane-associated protein 1-like, Anti-Stromal membrane-associated protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ASGMVAPMAMPAGYMGGMQASMMGVPNGMMTTQQAGYMAGMAAMPQTVYGVQPAQQLQWNLTQMTQQMAGMNFYGANGMMNYGQSMSGGNGQAANQTLSPQMWK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAP2(64744)

General description

The gene SMAP2 (stromal membrane-associated protein 2) is mapped to human chromosome 1p35.3-p34.1. It belongs to the SMAP subfamily of Arf (ADP-ribosylation factor) GTPase-activating proteins.

Immunogen

Stromal membrane-associated protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SMAP2 (stromal membrane-associated protein 2) regulates Arf (ADP-ribosylation factor) GTPase activity. It is important for transferrin endocytosis. SMAP2 interacts with another member of the family, SMAP1. It is associated with aspirin-intolerant asthma (AIA), a syndrome with lung inflammation and reversible bronchoconstriction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75466

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason Yongha Kim et al.
The Journal of asthma : official journal of the Association for the Care of Asthma, 47(9), 959-965 (2010-09-14)
Aspirin-intolerant asthma (AIA), as a clinical syndrome caused by aspirin, is characterized by lung inflammation and reversible bronchoconstriction. Recently, the altered trafficking and diminished airway reactivity have been implicated in allergic airway remodeling. The stromal membrane-associated protein 1-like (SMAP1L) exerts
Mami Sumiyoshi et al.
FEBS letters, 589(19 Pt B), 2754-2762 (2015-08-25)
In mammals, the small Arf GTPase-activating protein (SMAP) subfamily of Arf GTPase-activating proteins consists of closely related members, SMAP1 and SMAP2. These factors reportedly exert distinct functions in membrane trafficking, as manifested by different phenotypes seen in single knockout mice.
Ikuko Sakakura et al.
Biochemical and biophysical research communications, 404(2), 661-666 (2010-12-15)
Small G proteins play a central role in the organization of secretory and endocytotic pathways. The recruitment of some effectors, including vesicle coat proteins, is mediated by the ADP-ribosylation factor (Arf) family. Arf proteins have distinct subcellular localizations. ArfGAPs (Arf
Ilaria Del Giudice et al.
American journal of hematology, 89(1), 74-82 (2013-09-14)
Chronic lymphocytic leukemia (CLL) with stereotyped B-cell receptor (BCR) belonging to subset #1 (IGHV1-5-7/ IGKV1-39) display a poor outcome. To characterize their genetic and genomic features and BCR function, we selected 20 subset #1 CLL from a series of 579
Nobuhide Kobayashi et al.
Biochemical and biophysical research communications, 453(3), 473-479 (2014-10-05)
Arf GTPase-activating proteins (Arf GAP) play important roles in the formation of the membrane vesicles that traffic between subcellular membranous organelles. The small Arf GTPase-activating protein (SMAP) subfamily of Arf GAPs has two members, SMAP1 and SMAP2, in mammals. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service