Skip to Content
Merck
All Photos(3)

Key Documents

HPA021079

Sigma-Aldrich

Anti-PASK antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PAS domain-containing serine/threonine-protein kinase, Anti-PAS-kinase, Anti-PASKIN, Anti-hPASK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AQEVDVNSFSWNLKELFFSDQTDQTSSNCSCATSELRETPSSLAVGSDPDVGSLQEQGSCVLDDRELLLLTGTCVDLGQGRRFRESCVGHDPTEPLEVCLVSSEH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PASK(23178)

General description

The gene PASK (PAS domain-containing serine/threonine-protein kinase) is mapped to human chromosome 2q37. The protein is present in the cytoplasm as well as nucleus. PASK contains two PAS (Per-ARNT-Sim) domains and a kinase domain.

Immunogen

PAS domain-containing serine/threonine-protein kinase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PASK (PAS domain-containing serine/threonine-protein kinase) is a serine/threonine protein kinase, which controls insulin gene transcription by regulating PDX1 (pancreatic duodenal homeobox-1) phosphorylation. Presence of glucose induces the expression of PASK in pancreatic β cells. PASK also controls glycogen synthase and thereby is important for cellular energy homeostasis. PASK is also suggested to be involved in regulation of eukaryotic translation elongation factor eEF1A1 through phosphorylation. It is down-regulated in islets of humans suffering from type-2 diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73455

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Semplici et al.
The Journal of biological chemistry, 286(51), 44005-44014 (2011-11-09)
PAS kinase (PASK) is a glucose-regulated protein kinase involved in the control of pancreatic islet hormone release and insulin sensitivity. We aimed here to identify mutations in the PASK gene that may be associated with young-onset diabetes in humans. We
G da Silva Xavier et al.
Diabetologia, 54(4), 819-827 (2010-12-25)
We assessed whether per-arnt-sim (PAS) domain-containing protein kinase (PASK) is involved in the regulation of glucagon secretion. mRNA levels were measured in islets by quantitative PCR and in pancreatic beta cells obtained by laser capture microdissection. Glucose tolerance, plasma hormone
Meriem Semache et al.
The Journal of biological chemistry, 288(34), 24825-24833 (2013-07-16)
In pancreatic β-cells, glucose induces the binding of the transcription factor pancreatic duodenal homeobox-1 (PDX-1) to the insulin gene promoter to activate insulin gene transcription. At low glucose levels, glycogen synthase kinase 3β (GSK3β) is known to phosphorylate PDX-1 on
R An et al.
Biochemical Society transactions, 34(Pt 5), 791-793 (2006-10-21)
The transcription factor PDX-1 (pancreatic duodenal homeobox-1) is required for normal pancreatic development and for the function of insulin-producing islet beta-cells in mammals. We have shown previously that glucose regulates insulin gene expression in part through the activation and translocation
Gabriela da Silva Xavier et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(22), 8319-8324 (2004-05-19)
Per-Arnt-Sim (PAS) domain-containing kinases are common in prokaryotes, but a mammalian counterpart has only recently been described. Although the PAS domain of the mammalian PAS kinase (PASK) is closely related to the bacterial oxygen sensor FixL, it is unclear whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service