Skip to Content
Merck
All Photos(2)

Key Documents

HPA020361

Sigma-Aldrich

Anti-PRODH antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSPOX2, Anti-PIG6, Anti-PRODH1, Anti-PRODH2, Anti-Proline dehydrogenase (oxidase) 1, Anti-TP53I6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QESVAKLGIASRAEIEDWFTAETLGVSGTMDLLDWSSLIDSRTKLSKHLVVPNAQTGQLEPLLSRFTEEEELQMTRMLQRMDVLAKKATEMGVRLMVDAEQTYFQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRODH(5625)

General description

Proline dehydrogenase (oxidase) 1 (PRODH) is a mitochondrial enzyme expressed mainly in the brain. The gene encoding this enzyme is present on human chromosome 22q11.2 consisting of 14 exons.

Immunogen

proline dehydrogenase (oxidase) 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Proline dehydrogenase (oxidase) 1 (PRODH) catalyzes the first step in proline catabolism wherein it converts proline to D-1-pyrroline-5-carboxylate. It acts as a tumor suppressor, stimulates apoptosis and prevents proliferation of cells. PRODH also controls homeostasis and reactive oxygen species formation. The gene encoding this enzyme is deleted in 22q11 deletion syndrome (22q11DS) and mutations in it have been associated with neuropsychiatric disorders.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74554

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Suntsova et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(48), 19472-19477 (2013-11-13)
Using a systematic, whole-genome analysis of enhancer activity of human-specific endogenous retroviral inserts (hsERVs), we identified an element, hsERVPRODH, that acts as a tissue-specific enhancer for the PRODH gene, which is required for proper CNS functioning. PRODH is one of
Vanessa K Ota et al.
PloS one, 9(2), e87686-e87686 (2014-02-06)
Schizophrenia is a neurodevelopmental disorder with high heritability. Several lines of evidence indicate that the PRODH gene may be related to the disorder. Therefore, our study investigates the effects of 12 polymorphisms of PRODH on schizophrenia and its phenotypes. To
Shengdi Hu et al.
Experimental biology and medicine (Maywood, N.J.), 248(14), 1206-1220 (2023-08-26)
Glycine from sow's milk only meets 20% of the requirement of suckling piglets. However, how glycine is synthesized endogenously in neonates is not known. This study determined glycine synthesis from 4-hydroxyproline (an abundant amino acid in milk and neonatal blood)
Shengdi Hu et al.
Experimental biology and medicine (Maywood, N.J.), 248(17), 1446-1458 (2023-10-15)
This study tested the hypothesis that the synthesis of glycine from 4-hydroxyproline (an abundant amino acid in milk and neonatal blood) was impaired in tissues of piglets with intrauterine growth restriction (IUGR), thereby contributing to a severe glycine deficiency in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service