Skip to Content
Merck
All Photos(8)

Documents

HPA020265

Sigma-Aldrich

Anti-DBNL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cervical SH3P7, Anti-Cervical mucin-associated protein, Anti-Drebrin-F, Anti-Drebrin-like protein, Anti-HIP-55, Anti-HPK1-interacting protein of 55 kDa, Anti-SH3 domain-containing protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

INWTGEGVNDVRKGACASHVSTMASFLKGAHVTINARAEEDVEPECIMEKVAKASGANYSFHKESGRFQDVGPQAPVGSVYQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DBNL(28988)

General description

Drebrin-like protein (DBNL) is an adaptor protein with a C-terminal Src homology 3 (SH3) domain.

Immunogen

Drebrin-like protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Drebrin-like protein (DBNL) is involved in signal transduction pathways as it is a substrate for various kinases. It has important roles in endocytosis, immune responses and development of organs. It promotes cancer cell survival and inhibits apoptosis. DBNL also binds to 14-3-3 proteins and acts as a signal sensor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74993

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Liu et al.
International journal of molecular sciences, 15(3), 4903-4914 (2014-03-22)
In the present study, hematopoietic progenitor kinase 1 (HPK1)-interacting protein of 55 kDa (HIP-55) protein was over-expressed in HEK293 cells, which was genetically attached with 6x His tag. The protein was purified by nickel-charged resin and was then subjected to
Chengzhi Yang et al.
Molecular bioSystems, 10(6), 1393-1399 (2014-03-29)
HIP-55 (hematopoietic progenitor kinase 1 [HPK1]-interacting protein of 55 kDa) is the mammalian homologue of the yeast Abp1p. It contains a C-terminal Src homology 3 domain and an N-terminal actin depolymerization factor (ADF-H/C) domain. HIP-55 appears to be critical for
Drebrin-like protein DBN-1 is a sarcomere component that stabilizes actin filaments during muscle contraction
Eugenia Butkevich
Nature Communications, 7523 (2015)
Jin Han et al.
The Journal of biological chemistry, 278(52), 52195-52202 (2003-10-15)
HIP-55 (hematopoietic progenitor kinase 1 (HPK1)-interacting protein of 55 kDa, also called SH3P7 and mAbp1) is a novel SH3 domain-containing protein. HIP-55 binds to actin filaments both in vitro and in vivo. HIP-55 activates HPK1 and c-Jun N-terminal kinase (JNK)
Arindam Ghosh et al.
Biochemistry, 59(16), 1553-1558 (2020-04-14)
Drebrin-like protein (DBNL) is a multidomain F-actin-binding protein, which also interacts with other molecules within different intracellular pathways. Here, we present quantitative measurements on the size and conformation of human DBNL. Using dual-focus fluorescence correlation spectroscopy, we determined the hydrodynamic

Articles

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service