Skip to Content
Merck
All Photos(6)

Documents

HPA020044

Sigma-Aldrich

Anti-RAD21 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Double-strand-break repair protein rad21 homolog, Anti-NXP-1, Anti-Nuclear matrix protein 1, Anti-SCC1 homolog, Anti-hHR21

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

KLMMWKETGGVEKLFSLPAQPLWNNRLLKLFTRCLTPLVPEDLRKRRKGGEADNLDEFLKEFENPEVPREDQQQQHQQRDVIDEPIIEEPSRLQESVMEASRTNIDESAMPPPPPQGVKRKAGQIDPEPVMPPQQVEQMEIPPVEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD21(5885)

General description

RAD21 (RAD21 homolog, S. pombe) is a novel nuclear protein located on chromosome 8q24.

Immunogen

Double-strand-break repair protein rad21 homolog recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAD21 (RAD21 homolog, S. pombe) plays a crucial role in normal cell division. It acts as a main component of the cohesin complex for the proper chromosomal arrangement during cell division. The protein possesses two mitotic cleavage sites for the attachment of separase. It has been shown experimentally that at the beginning of anaphase stage, a caspase-like Esp1/separase attaches to the cleavage site of RAD21 and initiate sister chromatid separation, which is an essential step for the completion of cytokinesis. During double strand DNA damage repair, it helps the two DNA strands to adhere with each other. Mutation in RAD21 causes a congenital phenotype, cohesinopathy in humans characterized with growth retardation and minor skeletal anomalies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85184

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Chen et al.
The Journal of biological chemistry, 277(19), 16775-16781 (2002-03-05)
Caspases are a conserved family of proteases that play a critical role in the execution of apoptosis by cleaving key cellular proteins at Asp residues and modifying their function. Using an expression cloning strategy we recently developed, we isolated human
Kati P Porkka et al.
Genes, chromosomes & cancer, 39(1), 1-10 (2003-11-07)
To detect genes that are overexpressed in prostate cancer, a subtracted cDNA library was first constructed from the PC-3 cell line and subsequently screened by using cDNA microarray hybridization. Sixty-eight genes were found to be overexpressed (ratio>3) in PC-3. Half
Matthew A Deardorff et al.
American journal of human genetics, 90(6), 1014-1027 (2012-05-29)
The evolutionarily conserved cohesin complex was originally described for its role in regulating sister-chromatid cohesion during mitosis and meiosis. Cohesin and its regulatory proteins have been implicated in several human developmental disorders, including Cornelia de Lange (CdLS) and Roberts syndromes.
Debananda Pati et al.
Molecular and cellular biology, 22(23), 8267-8277 (2002-11-06)
Rad21 is one of the major cohesin subunits that holds sister chromatids together until anaphase, when proteolytic cleavage by separase, a caspase-like enzyme, allows chromosomal separation. We show that cleavage of human Rad21 (hRad21) also occurs during apoptosis induced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service