Skip to Content
Merck
All Photos(2)

Key Documents

HPA019191

Sigma-Aldrich

Anti-FGD5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-FYVE, RhoGEF and PH domain-containing protein 5, Anti-Zinc finger FYVE domain-containing protein 23

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

MLQHLNLDFHGAVMRALDDMDHEGRDTLAREELRQGLSELPAIHDLHQGILEELEERLSNWESQQKVADVFLAREQGFDHHATHILQFDRYLGLLSENCLHSPRLAAAVREFEQSVQGGSQTAKHRLLRVVQRLFQYQVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGD5(152273)

General description

The gene FGD5 (FYVE, RhoGEF and PH domain-containing protein 5) is mapped to human chromosome 3p25.1. It belongs to FGD family of guanine nucleotide exchange factors. In human umbilical vein endothelial cells, FGD5 is present at peripheral membrane ruffles and perinuclear regions. The protein contains a glutamate-rich amino-terminal region, a Dbl-homology (DH) domain, a first pleckstrin homology (PH) domain, an FYVE (Fab 1, YOTB, Vac 1 and EEA1) domain and a second carboxyl-terminal PH domain.

Immunogen

FYVE, RhoGEF and PH domain-containing protein 5 recombinant protein epitope signature tag (PrEST)

Application

Anti-FGD5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. The antibody has been used for immunocytochemistry in monolayer-cultured HUVECs (human umbilical vein endothelial cells).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Biochem/physiol Actions

Increase in levels of FGD5 (FYVE, RhoGEF and PH domain-containing protein 5) increases CDC42 (cell division cycle 42) activity. In vascular endothelial cells, FGD5 controls the proangiogenic action of VEGF (vascular endothelial growth factor). It modulates phosphatidylinositol 3-kinase signaling and thereby controls endothelial adhesion, survival and angiogenesis. FGD5 also participates in endothelial cell-targeted apoptosis and regulates vascular pruning.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72676

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Ando et al.
The Journal of cell biology, 202(6), 901-916 (2013-09-11)
Reorganization of the actin cytoskeleton is responsible for dynamic regulation of endothelial cell (EC) barrier function. Circumferential actin bundles (CAB) promote formation of linear adherens junctions (AJs) and tightening of EC junctions, whereas formation of radial stress fibers (RSF) connected
Marit Valla et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 66(11), 787-799 (2018-07-28)
Faciogenital dysplasia 5 ( FGD5) amplification drives tumor cell proliferation, and is present in 9.5% of breast cancers. We describe FGD5 expression, assess associations between FGD5 amplification and FGD5 expression, and assess FGD5 expression in relation to proliferation and prognosis.
Alexey A Dmitriev et al.
Epigenetics, 7(5), 502-513 (2012-04-12)
This study aimed to clarify genetic and epigenetic alterations that occur during lung carcinogenesis and to design perspective sets of newly identified biomarkers. The original method includes chromosome 3 specific NotI-microarrays containing 180 NotI clones associated with genes for hybridization
Maryam Nakhaei-Nejad et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(11), 2694-2701 (2012-08-28)
The function of the endothelial cell (EC)-enriched Rho family guanine nucleotide exchange factor, facio-genital dysplasia-5 (FGD5), is poorly understood. We sought to determine whether FGD5 regulates endothelial cytoskeletal reorganization and angiogenesis. We observed that FGD5 is expressed in primary human
Yusuke Kurogane et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(4), 988-996 (2012-02-14)
Vascular endothelial growth factor (VEGF) exerts proangiogenic action and induces activation of a variety of proangiogenic signaling pathways, including the Rho family small G proteins. However, regulators of the Rho family small G proteins in vascular endothelial cells (ECs) are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service