Skip to Content
Merck
All Photos(1)

Documents

HPA018317

Sigma-Aldrich

Anti-CALHM1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FAM26C antibody produced in rabbit, Anti-Protein FAM26C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

CTEHAKAFAKVCIQQFFEAMNHDLELGHTHGTLATAPASAAAPTTPDGAEEEREKLRGITDQGTMNRLLTSWHKCKPPLRLGQEEPPLMGNGW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALHM1(255022)

General description

The gene CALHM1 (calcium homeostasis modulator protein 1) is mapped to human chromosome 10q24.33. The gene encodes a multipass transmembrane glycoprotein. CALHM1 is present at the endoplasmic reticulum and plasma membrane. It has the ability to homomultimerize. CALHM1 is strongly expressed in adult brain and also in sweet/bitter/umami-sensing typeII taste bud cells.

Immunogen

Calcium homeostasis modulator protein 1 (Protein FAM26C)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Calcium homeostasis modulator protein 1 (CALHM1) controls cytosolic Ca2+ concentrations and amyloid β levels. CALHM1 polymorphism causing disturbance in Ca2+ and amyloid β levels is associated with Alzheimer′s disease. CALHM1 is also responsible for Ca2+ leakage from the endoplasmic reticulum (ER), thereby causing ER stress. CALHM1 plays crucial role in neuronal excitability. It positively controls the Ca2+-dependent activation of dual specificity mitogen-activated protein kinase kinase (MEK), extracellular signal-regulated kinase (ESK), ribosomal S6 kinase (RSK) and mitogen- and stress-activated protein kinase (MSK) signalling pathway in neurons. CALHM1 is also involved in release of taste-stimuli-evoked ATP from sweet-, bitter- and umami-sensing type II taste bud cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73092

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makiko Kashio et al.
Scientific reports, 9(1), 2681-2681 (2019-02-26)
The CALHM1/CALHM3 channel in the basolateral membrane of polarized taste cells mediates neurotransmitter release. However, mechanisms regulating its localization remain unexplored. Here, we identified CALHM1/CALHM3 in the basolateral membrane of type II taste cells in discrete puncta localized close to
Akiyuki Taruno
International journal of molecular sciences, 19(3) (2018-03-15)
Adenosine triphosphate (ATP) has been well established as an important extracellular ligand of autocrine signaling, intercellular communication, and neurotransmission with numerous physiological and pathophysiological roles. In addition to the classical exocytosis, non-vesicular mechanisms of cellular ATP release have been demonstrated
Ute Dreses-Werringloer et al.
Journal of cell science, 126(Pt 5), 1199-1206 (2013-01-25)
Calcium homeostasis modulator 1 (CALHM1) is a Ca(2+) channel controlling neuronal excitability and potentially involved in the pathogenesis of Alzheimer's disease (AD). Although strong evidence indicates that CALHM1 is required for neuronal electrical activity, its role in intracellular Ca(2+) signaling
Hafiz Sana-Ur-Rehman et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 312(5), R763-R772 (2017-03-04)
ATP release from urinary bladder is vital for afferent signaling. The aims of this study were to localize calcium homeostasis modulator 1 (CALHM1) and pannexin-1 expression and to determine their involvement in mediating ATP release in the bladder. To determine
Sonia Gallego-Sandín et al.
The Biochemical journal, 437(3), 469-475 (2011-05-18)
CALHM1 (calcium homoeostasis modulator 1), a membrane protein with similarity to NMDA (N-methyl-D-aspartate) receptor channels that localizes in the plasma membrane and the ER (endoplasmic reticulum) of neurons, has been shown to generate a plasma-membrane Ca(2+) conductance and has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service