Skip to Content
Merck
All Photos(3)

Key Documents

HPA018256

Sigma-Aldrich

Anti-NDRG3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein NDRG3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CAKGWIDWAASKLSGLTTNVVDIILAHHFGQEELQANLDLIQTYRMHIAQDINQDNLQLFLNSYNGRRDLEIERPILGQNDNKSKTLKCSTLLVVGD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDRG3(57446)

General description

The gene NDRG3 (N-myc downstream-regulated gene 3 protein) has been mapped to human chromosome 20q11.21-q11.23. The protein belongs to NDRG gene family. NDRG3 is expressed in many organs but is most abundant in brain, prostate and testis tissues. The protein is localized in the cytoplasm.

Immunogen

Protein NDRG3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

N-myc downstream-regulated gene 3 protein (NDRG3) is up-regulated in prostate cancer tissue and high expression correlates with advanced pathological stage and positive metastatic status. NDRG3 is up-regulated in hepatitis B virus (HBV)-related hepatocellular carcinoma cell line. NDRG3 is important for lactate-induced hypoxia signaling. Under normoxia, NDRG3 is degraded in a prolyl hydroxylase domain-containing protein-2 (PHD2)/von Hippel-Lindau disease tumor suppressor (VHL)-dependent manner. However, under hypoxia NDRG3 is protected by binding to lactate. The stable NDRG3 binds proto-oncogene c-Raf to mediate hypoxia-induced activation of Raf-ERK (extracellular signal-regulated kinase) pathway, promoting angiogenesis and cell growth. NDRG3 has been identified as a target for miR-122.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74470

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Chul Lee et al.
Cell, 161(3), 595-609 (2015-04-22)
Organisms must be able to respond to low oxygen in a number of homeostatic and pathological contexts. Regulation of hypoxic responses via the hypoxia-inducible factor (HIF) is well established, but evidence indicates that other, HIF-independent mechanisms are also involved. Here
R H Zhou et al.
Genomics, 73(1), 86-97 (2001-05-16)
RTP/Drg1/Cap43/rit42/TDD5/Ndr1/NDRG1 (referred to as NDRG1 hereafter) is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. Recently, the mutation of this gene was reported to be causative for hereditary motor and sensory neuropathy-Lom. Here, we cloned
Kyung Chan Park et al.
BMB reports, 48(6), 301-302 (2015-05-06)
Hypoxia is associated with many pathological conditions as well as the normal physiology of metazoans. We identified a lactate-dependent signaling pathway in hypoxia, mediated by the oxygen- and lactate-regulated protein NDRG family member 3 (NDRG3). Oxygen negatively regulates NDRG3 expression
Guo-feng Ren et al.
Histology and histopathology, 29(4), 535-542 (2013-11-14)
To investigate the clinicopathologic significance of NDRG2 and NDRG3, and their involvement in recurrence-free survival (RFS) and overall survival (OS) of prostate cancer (PCa). NDRG2 and NDRG3 expression in 206 pairs of primary PCa and corresponding noncancerous prostate tissue samples
Xianghu Qu et al.
Molecular and cellular biochemistry, 229(1-2), 35-44 (2002-04-09)
NDRG1 (N-Myc downstream regulated) is upregulated during cell differentiation, repressed by N-myc and c-myc in embryonic cells, and suppressed in several tumor cells. A nonsense mutation in the NDRG1 gene has been reported to be causative for hereditary motor and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service