Skip to Content
Merck
All Photos(1)

Documents

HPA015015

Sigma-Aldrich

Anti-SCYL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-N-terminal kinase-like protein, Anti-SCY1-like protein 1, Anti-Telomerase regulation-associated protein, Anti-Telomerase transcriptional element-interacting factor, Anti-Teratoma-associated tyrosine kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

AFPEDFCRHKVLPQLLTAFEFGNAGAVVLTPLFKVGKFLSAEEYQQKIIPVVVKMFSSTDRAMRIRLLQQMEQFIQYLDEPTVNTQIFPHVVHGFLDTNPAIREQTVKSMLLLAPKLNEANLNVELMK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCYL1(57410)

General description

SCYL1 (SCY1-like, kinase-like 1) belongs to the catalytically non-functional protein kinase family called Scy1-like protein family. It has a RKXX-COO- motif present at its C-terminal. It has a protein kinase-like domain at its N-terminal domain, and this protein is evolutionarily conserved. It is also called TEIF (telomerase transcriptional elements­interacting factor), acts as a transcription factor, and resides in centrosome. This gene is localized to human chromosome 11q13.

Immunogen

N-terminal kinase-like protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SCYL1 (SCY1-like, kinase-like 1) is involved in the COPI (coat protein)-mediated retrograde intracellular transport. It interacts with the appendage domain of COPG2 (coatomer subunit γ-2) through its RKXX-COO- sequence. Simultaneously, it interacts with Arf4, and other class II Arfs, and links them with COPG2-containing COPI complexes. Thus, it plays a key role in the organization of COPI coats, and acts as a scaffold for the same. In murine model, its inactivation results in motor neuron disease. It is a part of the nuclear tRNA export pathway, which is aminoacylation-dependent. It ensures the unloading of aminoacyl-tRNAs through nuclear pore complex, into the cytoplasm, from the nuclear tRNA export receptor. This protein is up-regulated in colorectal cancer (CRC), and this is related to the histological grade and the malignancy of CRC. This is also linked to centrosome amplification in CRC.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73500

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason N R Hamlin et al.
Journal of cell science, 127(Pt 7), 1454-1463 (2014-02-01)
Coatomer (COPI)-coated vesicles mediate membrane trafficking in the early secretory pathway. There are at least three subclasses of COPI coats and two classes of Arf GTPases that couple COPI coat proteins to membranes. Whether mechanisms exist to link specific Arfs
Shawn C Chafe et al.
Molecular biology of the cell, 21(14), 2483-2499 (2010-05-28)
Scyl1 is an evolutionarily conserved N-terminal protein kinase-like domain protein that plays a role in COP1-mediated retrograde protein trafficking in mammalian cells. Furthermore, loss of Scyl1 function has been shown to result in neurodegenerative disorders in mice. Here, we report
Ying Gao et al.
Ai zheng = Aizheng = Chinese journal of cancer, 28(12), 1277-1282 (2009-12-05)
Telomerase transcriptional elements-interacting factor (TEIF) gene found recently by our research group is a transcription factor of a kind of human telomerase reverse transcriptase (hTERT) gene, and expresses in many kinds of tumor tissues. This study was to evaluate the
Sebastien Gingras et al.
PloS one, 12(6), e0178680-e0178680 (2017-06-02)
A recent study identified SCYL1 as one of the components of the oncogenic STP axis, which promotes triple-negative breast cancer by regulating degradation of the REST tumor suppressor. Contrary to the findings of that study, herein we show by using
Genki Amano et al.
Molecular biology of the cell, 31(18), 1963-1973 (2020-06-26)
Arginine methylation is a common posttranslational modification that modulates protein function. SCY1-like pseudokinase 1 (SCYL1) is crucial for neuronal functions and interacts with γ2-COP to form coat protein complex I (COPI) vesicles that regulate Golgi morphology. However, the molecular mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service