Skip to Content
Merck
All Photos(2)

Key Documents

HPA014535

Sigma-Aldrich

Anti-ABCA10 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATP-binding cassette sub-family A member 10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

MYKVTRETHCWEFSPSMYFLSLEQIPKTPLTSLLIVNNTGSNIEDLVHSLKCQDIVLEIDDFRNRNGSDDPSYNGAIIVSGDQKDYRFSVACNTKKLNC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCA10(10349)

General description

ABCA10 (ATP-binding cassette, sub-family A (ABC1), member 10) belongs to ABC transporter superfamily. It is a part of the second group of ABCA subfamily, which contains five members located on chromosome 17q24. This protein is composed of 1543 amino acids, and is coded by a sequence of 4.6kb. This gene contains 40 exons and spans ~97kb. The transmembrane region of ABCA10 contains the N-terminal, and has a PEST sequence downstream to it. It is expressed ubiquitously, with predominant expression in heart, brain and the gastrointestinal (GI) tract.

Immunogen

ATP-binding cassette sub-family A member 10 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The functional characteristics of ABCA10 (ATP-binding cassette, sub-family A (ABC1), member 10) are yet unknown. Genome-wide association studies (GWAS) meta-analysis shows that variants of this gene might be linked to deaths related to follicular lymphoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72674

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fredrik Baecklund et al.
BMC medical genetics, 15, 113-113 (2014-10-09)
Survival in follicular lymphoma (FL) is highly variable, even within prognostic groups defined by tumor grade and the Follicular Lymphoma International Prognostic Index. Studies suggest that germline single nucleotide polymorphisms (SNPs) may hold prognostic information but further investigation is needed.
Jürgen J Wenzel et al.
Biochemical and biophysical research communications, 306(4), 1089-1098 (2003-06-25)
We recently identified several novel members of the A subclass of ABC transporters. In this study, we report the cloning of an additional ABC A subfamily transporter, denoted ABCA10, from macrophages. The coding sequence of ABCA10 is of 4.6 kb
M Dean et al.
Genome research, 11(7), 1156-1166 (2001-07-04)
The ATP-binding cassette (ABC) transporter superfamily contains membrane proteins that translocate a variety of substrates across extra- and intra-cellular membranes. Genetic variation in these genes is the cause of or contributor to a wide variety of human disorders with Mendelian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service