Skip to Content
Merck
All Photos(6)

Key Documents

HPA013411

Sigma-Aldrich

Anti-PDCD1LG2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-B7-DC, Anti-Butyrophilin B7-DC, Anti-CD273 antigen, Anti-PD-1-ligand 2, Anti-PD-L2, Anti-PDCD1 ligand 2, Anti-Programmed cell death 1 ligand 2 precursor, Anti-Programmed death ligand 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GYPLAEVSWPNVSVPANTSHSRTPEGLYQVTSVLRLKPPPGRNFSCVFWNTHVRELTLASIDLQSQMEPRTHPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Programmed cell death 1 ligand 2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PDCD1LG2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PDCD1LG2 (Programmed cell death 1 ligand 2) gene encodes a ligand for PD-1 that serves as a negative regulator of T cell activation. The encoded protein is also called as PD-L2 and is induced on inflammatory macrophages. It is expressed mainly on dendritic cells and the expression is induced by IFN-γ, granulocyte/macrophage colony-stimulating factor, and IL-4. It functions in T-cell proliferation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72423

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dengbo Ji et al.
Cancer immunology research, 6(11), 1401-1416 (2018-10-05)
Checkpoint blockade therapy triggers tumor-specific immune responses in a variety of cancer types. We presumed that rectal cancer patients could have become sensitive to immunotherapy after receiving neoadjuvant chemoradiotherapy (nCRT). In this study, we report immune alternation in post-nCRT patients
Mengyuan Shi et al.
Cancers, 15(9) (2023-05-13)
Colorectal cancer (CRC) is the third most common type of cancer. The ultraviolet radiation resistance-associated gene (UVRAG) plays a role in autophagy and has been implicated in tumor progression and prognosis. However, the role of UVRAG expression in CRC has
P'ng Loke et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(9), 5336-5341 (2003-04-17)
PD-L1 and PD-L2 are ligands for PD-1, a costimulatory molecule that plays an inhibitory role in regulating T cell activation in the periphery. We find that PD-L1 is highly expressed on inflammatory macrophages as compared with resident peritoneal macrophages but
Alexander C Dowell et al.
Frontiers in oncology, 11, 626748-626748 (2021-03-16)
The use of immune checkpoint blockade, in particular PD-1 and PD-L1 inhibitors, is now commonplace in many clinical settings including the treatment of muscle-invasive bladder cancer (MIBC). Notwithstanding, little information exists regarding the expression of the alternative PD-1 ligand, PD-L2
Yang Zhang et al.
OncoTargets and therapy, 7, 567-573 (2014-04-22)
The clinicopathologic characteristics of tumors expressing programmed death (PD-1) ligands (PD-Ls) PD-L1 or PD-L2 and their associations with common driver mutations in lung adenocarcinoma are not clearly defined, despite the progression of anti-PD-1/PD-L1 immunotherapy. PD-L1 and PD-L2 expression was measured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service