Skip to Content
Merck
All Photos(1)

Documents

HPA012036

Sigma-Aldrich

Anti-PANK4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Pantothenate kinase 4, Anti-Pantothenic acid kinase 4, Anti-hPanK4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

FDHSGKDTEREHEPPYEISVQEEITARLHFIKFENTYIEACLDFIKDHLVNTETKVIQATGGGAYKFKDLIEEKLRLKVDKEDVMTCLIKGCNFVLKNIPHEAFVYQKDSDPEFRFQTNHPHIFPYLLVNIGSGVSIVKVETE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PANK4(55229)

Immunogen

Pantothenate kinase 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PANK4 (Pantothenate kinase 4) is an essential rate-controlling enzyme involved in several synthetic pathways. It maintains the flux balance in cardiac muscle. The functional activity of the gene is inhibited by almost all the acyl esters including coenzyme A in both liver and heart. However, free carnitine activates the PANK4. Mutation in PANK4 causes an autosomal recessive neurodegenerative disorder, hallervorden-Spatz syndrome (HSS) characterized with extrapyramidal dysfunction, onset in childhood, and a relentlessly progressive course. The gene alteration changes the iron accumulation in the basal ganglia of brain. It has been reported that PanK4 may play an important role in the glucose metabolism via controlling the M2-type pyruvate kinase (Pkm2) activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70337

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunfeng Li et al.
Molecular and cellular biochemistry, 277(1-2), 117-125 (2005-09-01)
A new Rattus norvegicus PanK gene was isolated by mRNA differential display from high concentration glucose-stimulated rat, which encodes a human PanK4-like protein with KOG2201 and KOG4584 domain. Proteins that interact with rat PanK4 were identified by the application of
B Zhou et al.
Nature genetics, 28(4), 345-349 (2001-08-02)
Hallervorden-Spatz syndrome (HSS) is an autosomal recessive neurodegenerative disorder associated with iron accumulation in the brain. Clinical features include extrapyramidal dysfunction, onset in childhood, and a relentlessly progressive course. Histologic study reveals iron deposits in the basal ganglia. In this
J D Robishaw et al.
The American journal of physiology, 248(1 Pt 1), E1-E9 (1985-01-01)
The metabolism of coenzyme A and control of its synthesis are reviewed. Pantothenate kinase is an important rate-controlling enzyme in the synthetic pathway of all tissues studied and appears to catalyze the flux-generating reaction of the pathway in cardiac muscle.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service